Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HA-1 Mouse anti-Nonspecies specific, Clone: 12CA5, Novus Biologicals™

Mouse Monoclonal Antibody
$231.50 - $499.50
Specifications
Antigen | HA-1 |
---|---|
Clone | 12CA5 |
Dilution | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
HA-1 Monoclonal antibody specifically detects HA-1 in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
HA-1 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
Non-species specific | |
histocompatibility (minor) HA-1, HLA-HA1, KIAA0223HA-1, minor histocompatibility antigen HA-1, minor histocompatibility protein HA-1 | |
Balb/C mice were immunized with a 9 amino acid petide to influenza virus hemagglutinin (Uniprot: Q3TBD2) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
12CA5 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Signal Transduction | |
PBS | |
23526 | |
IgG2b | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title