Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ HA Synthetic Peptide PROMO
Click to view available options
:
Thermo Scientific™ HA Synthetic Peptide
Description
The HA peptide, NH2-YPYDVPDYA-COOH, is a synthetic peptide used to competitively elute fusion proteins containing the influenza hemagglutinin (HA) tag bound to immobilized anti-HA affinity resins.
Formulation: 5 mg purified Peptide (1mg/mL) no preservative
- 26184 detects HA in samples. 26184 has been successfully used in Control procedures.
Specifications
Specifications
| Protein Form | Recombinant |
| Content And Storage | Store at 4°C. |
| For Use With (Application) | Immunoprecipitation |
| Quantity | 5 mg |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction