Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Antibody (12CA5[2), Novus Biologicals™
SDP

Catalog No. NB018663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018663 0.05 mg
NBP150701 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018663 Supplier Novus Biologicals Supplier No. NBP1507010.05MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HA Tag Monoclonal specifically detects HA Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen HA Tag
Applications Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 12CA5[2]
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL, Immunocytochemistry/Immunofluorescence 10 - 20 ug/mL, Immunoprecipitation 10 - 20 ug/mL
Formulation PBS (pH 7.2), 50% glycerol, 1% BSA with 0.02% Thimerosal
Gene Alias HA epitope tag, Hemagglutinin
Host Species Mouse
Immunogen Synthetic peptide (YPYDVPDYA) derived from the human influenza hemaglutinin protein
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.