Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HA Tag Antibody (16.43), PE/Atto594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250416PEATT594
Description
HA Tag Monoclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Flow Cytometry.Specifications
HA Tag | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
Rat | |
Protein G purified | |
Epitope Tags, HA Epitope Tags | |
Store at 4°C in the dark. Do not freeze. | |
Epitope Tag |
Flow Cytometry | |
16.43 | |
Flow Cytometry | |
HA epitope tag, Hemagglutinin | |
This HA Tag Antibody (16.43) was developed against the C-terminal influenza virus hemagglutinin (HA) tag, YPYDVPDYA. | |
0.1 mL | |
Primary | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction