Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HA Tag Antibody, Alexa Fluor™ 700, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB600362AF700
Description
HA Tag Polyclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.Specifications
HA Tag | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
HA epitope tag, Hemagglutinin | |
This HA Tag Antibody was developed by immunizing goats with HA cleavage site (YPYDVPDYA) conjugated to KLH. | |
0.1 mL | |
Epitope Tags, HA Epitope Tags | |
Store at 4C in the dark. | |
Epitope Tag |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction