Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HA Tag Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $561.00
Specifications
Antigen | HA Tag |
---|---|
Clone | 912426 |
Dilution | Western Blot 0.5 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Description
HA Tag Monoclonal specifically detects HA Tag in Multi-Species samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
HA Tag | |
Western Blot 0.5 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HA, HA epitope tag, Hemagglutinin | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
912426 | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Multi-species | |
ABB51961 | |
HA Tag peptide Accession # ABB51961 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects N-terminal,C-terminal, and Internal Sequence HA-tagged proteins in Western blot. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title