Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HA Tag Monoclonal Antibody (2-2.2.14), DyLight™ 680
GREENER_CHOICE

Catalog No. PI26183D680
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26183D680 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26183D680 Supplier Invitrogen™ Supplier No. 26183D680
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

26183-D680 has been successfully used in immunofluorescence and Western blot applications. DyLight 680 has an excitation/emission of 692/712 nm. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.

HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HA Tag
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2-2.2.14
Concentration 1 mg/mL
Conjugate DyLight 680
Formulation PBS with proprietary stabilizer and 0.02% sodium azide
Gene Alias HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin
Host Species Mouse
Immunogen HA peptide YPYDVPDYA derivitized to ovalbumin.
Purification Method Protein G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.