Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Monoclonal Antibody (7H5), Bioss
SDP

Catalog No. 502280641
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0641 Supplier Bioss Supplier No. BSM50131M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HA Tag Monoclonal antibody specifically detects HA Tag in Tag samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen HA Tag
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7H5
Conjugate Unconjugated
Formulation HEPES with 1% BSA, 50% glycerol and 0.03% sodium azide
Gene Alias HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin
Host Species Mouse
Immunogen Recombinant Human HA-tag protein purified E.coli
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.