Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag mouse anti-Tag, Magnetic beads, Clone: CB051, liquid, TrueMAB™
SDP

Catalog No. 501671200
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1200 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1200 Supplier OriGene Supplier No. TA183070
Add to Cart
Add to Cart

mouse Monoclonal Antibody

Synthetic peptide sequence is YPYDVPDYA

HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.

Specifications

Antigen HA Tag
Applications Immunoprecipitation
Classification Monoclonal
Clone CB051
Concentration 1 mg/mL
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Alias Human influenza hemagglutinin; hemagglutinin; HA-tag; HA; HA Epitope Tag
Host Species Mouse
Immunogen Monoclonal antibody is produced by immunizing mice with a synthetic peptide from influenza hemagglutinin coupled to KLH
Purification Method Affinity Chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.