Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA-Tag Rabbit anti-Human, Biotin, Clone: RM305, Abnova™

Catalog No. 89339228 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-228 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-228 Supplier Abnova Corporation Supplier No. MAB22012
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

HA-Tag Monoclonal antibody specifically detects HA-Tag in Human samples. It is validated for IP, ICC, IHC-P, WB-Tr, Flow Cyt.

Specifications

Antigen HA-Tag
Applications Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry
Classification Monoclonal
Clone RM305
Conjugate Biotin
Host Species Rabbit
Immunogen A peptide corresponding to amino acids 98-106 of HA-tag
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.