Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HA Tag Recombinant Rabbit Monoclonal Antibody (SN07-06)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542442
This item is not returnable.
View return policy
Description
Positive Control: HA tag recombinant protein, C-terminal HA-tagged recombinant protein, N-terminal HA-tagged recombinant protein.
HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
Specifications
| HA Tag | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Tag | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| SN07-06 | |
| Unconjugated | |
| HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
| Recombinant protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Tag |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction