Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAAO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581604 Supplier Invitrogen™ Supplier No. PA581604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat HAAO/3-HAO.

HAAO (3-Hydroxyanthranilate 3, 4-dioxygenase) is a monomeric cytosolic protein of the family of intramolecular dioxygenases containing non-heme ferrous iron. It is widely distributed in peripheral organs, such as liver and kidney, and is present in low amounts in the central nervous system. This enzyme participates in tryptophan metabolism. It employs one cofactor, iron. HAAO catalyzes the synthesis of quinolinic acid (QUIN) from 3-hydroxyanthranilic acid. QUIN is an excitotoxin whose toxicity is mediated by its ability to activate glutamate N-methyl-D-aspartate receptors. Increased cerebral levels of QUIN may participate in the pathogenesis of neurological and inflammatory disorders. HAAO has been suggested to play a role in disorders associated with altered tissue levels of QUIN. Furthermore, recent study shows that HAAO are excellent candidate biomarkers for detecting ovarian cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAAO
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HAAO
Gene Accession No. P46953
Gene Alias 0610007K21Rik; 0610012J07Rik; 3HAO; 3-HAO; 3-HAO {ECO:0000255; 3-HAOxase; 3-hydroxyanthranilate 3,4-dioxygenase; 3-hydroxyanthranilate 3,4-dioxygenase {ECO:0000255; 3-hydroxyanthranilate oxygenase; 3-hydroxyanthranilate oxygenase {ECO:0000255; 3-hydroxyanthranilic acid dioxygenase; 3-hydroxyanthranilic acid dioxygenase {ECO:0000255; Haao; HAD; HAD {ECO:0000255; HAMAP-Rule:MF_03019}; HAO
Gene Symbols HAAO
Host Species Rabbit
Immunogen Recombinant Rat HAAO/3-HAO Protein (Met1-Gly286).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56823
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.