Learn More
IKA™ Habitat Cell Photobioreactor Control Tower
SDP
Optimized photobioreactor for cell culture applications with LED panels
Specifications
For Use With (Equipment) Photosynthetic Organisms
View More Specs
Products 2
Catalog Number Mfr. No. Product Line Price Quantity & Availability  
Catalog Number Mfr. No. Product Line Price Quantity & Availability  
11-116-972
SDP
View Documents
IKA™
0010007554
Cell Photobioreactor N/A N/A
Please contact your sales representative for a quote or further ordering instructions.
11-116-974
SDP
View Documents
IKA™
0010007596
Ferment Photobioreactor N/A N/A
Please contact your sales representative for a quote or further ordering instructions.
Description
Description
  • Ideal for applications involving photosynthetic organisms, such as algae cultivation.
  • Latest technology sensors for better insights and more control
  • Intuitive, easy-to-use control software that provides simple initial setup and intuitive user assistance throughout the process
  • Includes LED panels essential for transforming HABITAT research bioreactor into a photobioreactor
  • Four integrated Watson-Marlow pumps with adjustable direction and speed offer highest precision and versatility in pumping acids, bases, anti-foam agents, and nutrient solutions for the management of media in photobioreactors and fermentation processes
  • High precision motor speed control
  • Integrated PID control ensures precise temperature regulation at all times
  • TUV Certified
Specifications
Specifications
Photosynthetic Organisms
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.