Description
- Fermentation vessel packages are equipped with a ring sparger and 6-bladed propeller stirrer
- High gas flow rate of 0 to 20,000 cc/min for fermentation
- Additional sensors are available upon request
- Borosilicate glass reactor material
- Stainless steel 1.4404 lid material
- 18 available lid ports
Scope of Delivery:
- Glass vessel
- Vessel holder
- Lid complete
- Driving shaft
- Motor coupling
- Ring sparger
- Condenser
- Baffle
- 6 Blade segment impeller
- Temp. sensor
- pH sensor
- DO sensor
- Foam sensor
- Level sensor
- Heating blanket
- Cable and tube set
- 4 x Sample flask 250 mL
- Syringe set
Specifications
Specifications
| Product Type | Bioreactor Package |
| Autoclavable | Autoclavable |
| Flow Rate | 20000 cc/min |
| Material | Borosilicate glass |