Learn More
IKA™ Habitat Ferment Photobioreactor Control Tower
Connectivity options for gas, liquid addition, sensors and temperature control for seamless operation
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
11-116-973
![]() |
IKA™
0010007564 |
Control Tower | N/A | N/A | |||||
Please contact your sales representative for a quote or further ordering instructions. | |||||||||
11-116-975
![]() |
IKA™
0010007606 |
Control Tower CCT | N/A | N/A | |||||
Please contact your sales representative for a quote or further ordering instructions. | |||||||||
Description
- Ideal for applications involving photosynthetic organisms, such as algae cultivation.
- Intuitive, easy-to-use control software that provides simple initial setup and intuitive user assistance throughout the process
- Includes LED panels essential for transforming HABITAT research bioreactor into a photobioreactor
- Four integrated Watson-Marlow pumps with adjustable direction and speed offer highest precision and versatility in pumping acids, bases, anti-foam agents, and nutrient solutions for the management of media in photobioreactors and fermentation processes
- High precision motor speed control
- Cross platform connectivity: USB, PC, RS232, OPC-UA, ethernet, external signal input, external pump, single-use unit, thermostat
- Status LED display direct error display by indicator light
- Integrated PID control ensures precise temperature regulation at all times
Specifications
Ferment Photobioreactor |