Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HABP1/C1QBP/GC1q R Antibody (1G7), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242691PEATT594
Description
HABP1/C1QBP/GC1q R Monoclonal specifically detects HABP1/C1QBP/GC1q R in Human samples. It is validated for Flow Cytometry.Specifications
HABP1/C1QBP/GC1q R | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
C1QBP | |
Recombinant human C1QBP (74-282aa) purified from E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
1G7 | |
Flow Cytometry | |
C1q globular domain-binding protein, C1qBP, complement component 1 Q subcomponent-binding protein, mitochondrial, complement component 1, q subcomponent binding protein, GC1QBP, gC1qR, gC1Q-R, GC1q-R protein, Glycoprotein gC1qBP, HABP1p33, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, p32, SF2P32, splicing factor SF2-associated protein | |
Mouse | |
Protein A purified | |
Stem Cell Markers | |
708.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction