Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HABP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA554009
Description
Immunogen sequence: GHCALESKYV KTVCLPDGSF PSGSECHISG WGVTETGKGS RQLLDAKVKL IANTLCNSRQ LYDHMIDDSM ICAGNLQKPG QDTCQGDSGG PLTCEKDGTY YVYGI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
HABP2 Polyclonal antibody specifically detects HABP2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| HABP2 | |
| Polyclonal | |
| Unconjugated | |
| HABP2 | |
| AI035669; Factor seven-activating protease; factor VII activating protein; factor VII-activating protease; FSAP; HABP; Habp2; Hepatocyte growth factor activator-like protein; HGFAL; hyaduronic acid-binding protein 2; hyaluronan binding protein 2; hyaluronan-binding protein 2; Hyaluronan-binding protein 2 27 kDa light chain; Hyaluronan-binding protein 2 27 kDa light chain alternate form; Hyaluronan-binding protein 2 50 kDa heavy chain; Hyaluronan-binding protein 2 50 kDa heavy chain alternate form; hyaluronic acid binding protein 2; NMTC5; PHBP; plasma hyalurona; plasma hyaluronan binding protein; plasma hyaluronan-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3026 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14520 | |
| HABP2 | |
| Recombinant protein corresponding to Human HABP2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction