Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HABP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563225
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563225 Supplier Invitrogen™ Supplier No. PA563225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGGPGNRVFD AFDQRGKREF ERYGGNDKIA VRTEDNMGGC GVRTWGSGKD TSDVEPTAPM EEPTVVEESQ GTPEEESPAK VPE Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 75%.

RNA-binding protein that plays a role in the regulation of transcription, pre-mRNA splicing and mRNA translation (PubMed:14699138, PubMed:16455055, PubMed:19523114, PubMed:21771594). Negatively regulates DNA-binding activity of the transcription factor MEF2C in myocardial cells in response to mechanical stress. Plays a role in pre-mRNA splicing regulation (PubMed:19523114). Binds (via C-terminus) to poly(U) RNA (PubMed:19523114). Involved in mRNA translation regulation, probably at the initiation step (PubMed:21771594). Seems to play a role in PML-nuclear bodies formation (PubMed:28695742). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HABP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HABP4
Gene Accession No. Q5JVS0
Gene Alias 4933413D03Rik; 4933428J01Rik; chromodomain helicase DNA binding protein 3 interacting protein; habp4; hyaluronan binding protein 4; Hyaluronan-binding protein 4; hyaluronic acid binding protein 4; Hyaluronic acid-binding protein 4; hyaluronic acid-binding protein 4 (intracellular); IHABP4; IHABP-4; intracellular antigen detected by monoclonal antibody Ki-1; intracellular hyaluronan-binding protein 4; intracellular hyaluronan-binding protein p57; Ki-1/57; ki-1/57 intracellular antigen; SERBP1L
Gene Symbols HABP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HABP4. Recombinant protein control fragment (Product #RP-101784).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22927
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.