Learn More
Invitrogen™ HABP4 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | HABP4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | HABP4 |
| Gene Accession No. | Q5JVS0 |
| Gene Alias | 4933413D03Rik; 4933428J01Rik; chromodomain helicase DNA binding protein 3 interacting protein; habp4; hyaluronan binding protein 4; Hyaluronan-binding protein 4; hyaluronic acid binding protein 4; Hyaluronic acid-binding protein 4; hyaluronic acid-binding protein 4 (intracellular); IHABP4; IHABP-4; intracellular antigen detected by monoclonal antibody Ki-1; intracellular hyaluronan-binding protein 4; intracellular hyaluronan-binding protein p57; Ki-1/57; ki-1/57 intracellular antigen; SERBP1L |
| Gene Symbols | HABP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.