Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HABP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140105
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LEVEEETQVQ EMTLDEWKNL QEQTRPKPEF NIRKPESTVP SKAVVIHKSK YRDDMVKDDY EDDSHVFR.
RNA-binding protein that plays a role in the regulation of transcription, pre-mRNA splicing and mRNA translation (PubMed:14699138, PubMed:16455055, PubMed:19523114, PubMed:21771594). Negatively regulates DNA-binding activity of the transcription factor MEF2C in myocardial cells in response to mechanical stress. Plays a role in pre-mRNA splicing regulation (PubMed:19523114). Binds (via C-terminus) to poly(U) RNA (PubMed:19523114). Involved in mRNA translation regulation, probably at the initiation step (PubMed:21771594). Seems to play a role in PML-nuclear bodies formation (PubMed:28695742). [UniProt].
Specifications
HABP4 | |
Polyclonal | |
Unconjugated | |
HABP4 | |
4933413D03Rik; 4933428J01Rik; chromodomain helicase DNA binding protein 3 interacting protein; habp4; hyaluronan binding protein 4; Hyaluronan-binding protein 4; hyaluronic acid binding protein 4; Hyaluronic acid-binding protein 4; hyaluronic acid-binding protein 4 (intracellular); IHABP4; IHABP-4; intracellular antigen detected by monoclonal antibody Ki-1; intracellular hyaluronan-binding protein 4; intracellular hyaluronan-binding protein p57; Ki-1/57; ki-1/57 intracellular antigen; SERBP1L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22927 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JVS0 | |
HABP4 | |
Recombinant protein corresponding to Human HABP4. Recombinant protein control fragment (Product #RP-109019). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction