Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HABP4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15402R

Description
RNA-binding protein that plays a role in the regulation of transcription, pre-mRNA splicing and mRNA translation. Negatively regulates DNA-binding activity of the transcription factor MEF2C in myocardial cells in response to mechanical stress. Plays a role in pre-mRNA splicing regulation. Binds (via C-terminus) to poly(U) RNA. Involved in mRNA translation regulation, probably at the initiation step. Seems to play a role in PML-nuclear bodies formation.Specifications
HABP4 | |
Polyclonal | |
Unconjugated | |
HABP4 | |
4933413D03Rik; 4933428J01Rik; BOS_9190; chromodomain helicase DNA binding protein 3 interacting protein; Habp4; hyaluronan binding protein 4; Hyaluronan-binding protein 4; hyaluronic acid binding protein 4; Hyaluronic acid-binding protein 4; hyaluronic acid-binding protein 4 (intracellular); IHABP4; IHABP-4; intracellular antigen detected by monoclonal antibody Ki-1; Intracellular hyaluronan-binding protein 4; intracellular hyaluronan-binding protein p57; Ki-1/57; Ki-1/57 intracellular antigen; SERBP1L | |
Rabbit | |
Protein A | |
RUO | |
22927 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5JVS0 | |
HABP4 | |
KLH conjugated synthetic peptide derived from human HABP4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction