Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HACE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583803 Supplier Invitrogen™ Supplier No. PA583803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVNENDILLV HRDSIFRSSC EVVSKANCAK LKQGIAVRFH GEEGMGQGVV REWFDILSNE IVNPDYALFT QSADGTT Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.

E3 ubiquitin-protein ligase involved in Golgi membrane fusion and regulation of small GTPases. Acts as a regulator of Golgi membrane dynamics during the cell cycle: recruited to Golgi membrane by Rab proteins and regulates postmitotic Golgi membrane fusion. Acts by mediating ubiquitination during mitotic Golgi disassembly, ubiquitination serving as a signal for Golgi reassembly later, after cell division. Specifically interacts with GTP-bound RAC1, mediating ubiquitination and subsequent degradation of active RAC1, thereby playing a role in host defense against pathogens. May also act as a transcription regulator via its interaction with RARB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HACE1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HACE1
Gene Accession No. Q8IYU2
Gene Alias 1700042J16Rik; A730034A22Rik; BC025474; E3 ubiquitin-protein ligase HACE1; hace 1; Hace1; HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat-containing E3 ubiquitin-protein ligase 1; HECT-type E3 ubiquitin transferase HACE1; KIAA1320; SPPRS
Gene Symbols HACE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HACE1. Recombinant protein control fragment (Product #RP-106106).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.