Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HACE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30390420UL
Description
HACE1 Polyclonal specifically detects HACE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HACE1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
57531 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
E3 ubiquitin-protein ligase HACE1, EC 6.3.2, HACE, HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1, HECT domain and ankyrin repeat-containing E3 ubiquitin-protein ligase 1, KIAA1320EC 6.3.2.- | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HACE1 (NP_065822.2). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction