Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HACL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557295 Supplier Invitrogen™ Supplier No. PA557295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVEGDSAFGF SGMEVETICR YNLPIILLVV NNNGIYQGFD TDTWKEMLKF QDATAVVPPM CLLPNSHYEQ VMTAFGGKGY FVQTPEELQ Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

HACL1 gene ontology annotations related to this gene include lyase activity; magnesium ion binding; thiamine pyrophosphate binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HACL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hacl1
Gene Accession No. Q8CHM7, Q9QXE0, Q9UJ83
Gene Alias 1600020H07Rik; 2hpcl; 2-HPCL; 2-hydroxyacyl-CoA lyase 1; 2-hydroxyphytanol-CoA lyase; 2-hydroxyphytanoyl-CoA lyase; 2-hydroxyphytanoyl-Coenzyme A lyase; Hacl1; Hpcl; Hpcl2; HSPC279; PHYH2; Phytanoyl-CoA 2-hydroxylase 2; phytanoyl-CoA hydroxylase 2; phytanoyl-Coenzyme A 2-hydroxylase 2
Gene Symbols Hacl1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HACL1. Recombinant protein control fragment (Product #RP-96331).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26061, 56794, 85255
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.