Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HADA
GREENER_CHOICESDP

Catalog No. 66475 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-475 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 66-475 Supplier Tocris Bioscience™ Supplier No. 6647/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fluorescent Probes for Imaging Bacteria

HADA is a fluorescent D-amino acid. Suitable for labeling peptidoglycans in live bacteria. Results in strong peripheral and septal labeling of diverse bacterial cell populations without affecting growth rates. Displays mitochondrial outer membrane permeability. Excitation/emission λ ∽405/450 nm.

Specifications

Chemical Name or Material 3-[[(7-Hydroxy-2-oxo-2H-1-benzopyran-3-yl)carbonyl]amino]-D-alanine hydrocholoride
CAS 2253733-10-5
Quantity 5 mg
Molecular Formula C13H12N2O6.HCl
Formula Weight 328.71
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.