Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HADH Monoclonal Antibody (1A12BC8)
GREENER_CHOICE

Catalog No. A21979
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21979 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21979 Supplier Invitrogen™ Supplier No. A21979
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HADH Monoclonal Antibody for ICC/IF, IHC (P), IP, ELISA

This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene of this gene on chromosome 15.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HADH
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 1A12BC8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene HADH
Gene Accession No. Q16836
Gene Alias AA409008; AU019341; AW742602; Had; HADH; HADH1; HADHSC; HCDH; HHF4; hydroxyacyl-CoA dehydrogenase; hydroxyacyl-Coenzyme A dehydrogenase; hydroxyacyl-coenzyme A dehydrogenase, mitochondrial; hydroxylacyl-Coenzyme A dehydrogenase short chain; hydroxylacyl-Coenzyme A dehydrogenase, short chain; hydroxylacyl-Coenzyme A dehydrogenase-dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain; M/SCHAD; medium and short chain L-3-hydroxyacyl-coenzyme A dehydrogenase; medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase; MGC8392; Mschad; SCHAD; short chain 3-hydroxyacyl-CoA dehydrogenase; short-chain 3-hydroxyacyl-CoA dehydrogenase; testis secretory sperm-binding protein Li 203a
Gene Symbols HADH
Host Species Mouse
Immunogen Human liver mitochondria.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3033
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.