Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HADHA Mouse anti-Human, Mouse, Clone: 3E9B1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | HADHA |
|---|---|
| Clone | 3E9B1 |
| Concentration | 1 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6347
![]()
|
Proteintech Group Inc
602501IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes the alpha subunit of the mitochondrial trifunctional protein, which catalyzes the last three steps of mitochondrial beta-oxidation of long chain fatty acids. The mitochondrial membrane-bound heterocomplex is composed of four alpha and four beta subunits, with the alpha subunit catalyzing the 3-hydroxyacyl-CoA dehydrogenase and enoyl-CoA hydratase activities. Mutations in this gene result in trifunctional protein deficiency or LCHAD deficiency. The genes of the alpha and beta subunits of the mitochondrial trifunctional protein are located adjacent to each other in the human genome in a head-to-head orientation.Specifications
| HADHA | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 78 kDa gastrin binding protein, ECHA, HADH, HADHA, LCHAD, MTPA, TP ALPHA | |
| Hadha | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3E9B1 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P40939, Q8BMS1 | |
| 3030, 97212 | |
| HADHA Fusion Protein Ag1211 | |
| Primary | |
| -20°C | |
| HADHA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title