Learn More
Description
Specifications
Specifications
| Purity | Greater than 80% by SDS-PAGE and Coomassie blue staining |
| Common Name | HADHA Recombinant Protein Antigen |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Antibody Competition |
| Molecular Weight (g/mol) | 28 kDa |
| Product Type | Recombinant Protein Antigen |
| Quantity | 100 μL |
| Sequence | DEDIQFRLVTRFVNEAVMCLQEGILATPAEGDIGAVFGLGFPPCLGGPFRFVDLYGAQKIVDRLKKYEAAYGKQFTPCQLLADHANSPNKK |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-50314. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
