Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HADHB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583376 Supplier Invitrogen™ Supplier No. PA583376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRDFMYVSQD PKDQLLLGPT YATPKVLEKA GLTMNDIDAF EFHEAFSGQI LANFKAMDSD WFAENYMGRK TKVGLPPLEK FNNWGGSLSL G Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.

This gene encodes the beta subunit of the mitochondrial trifunctional protein, which catalyzes the last three steps of mitochondrial beta-oxidation of long chain fatty acids. The mitochondrial membrane-bound heterocomplex is composed of four alpha and four beta subunits, with the beta subunit catalyzing the 3-ketoacyl-CoA thiolase activity. The encoded protein can also bind RNA and decreases the stability of some mRNAs. The genes of the alpha and beta subunits of the mitochondrial trifunctional protein are located adjacent to each other in the human genome in a head-to-head orientation. Mutations in this gene result in trifunctional protein deficiency. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HADHB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HADHB
Gene Accession No. P55084, Q60587, Q99JY0
Gene Alias 2-enoyl-Coenzyme A (CoA) hydratase, beta subunit; 3-ketoacyl-CoA thiolase; 3-ketoacyl-Coenzyme A (CoA) thiolase of mitochondrial trifunctional protein, beta subunit; 4930479F15Rik; acetyl-CoA acyltransferase; bet; Beta-ketothiolase; ECHB; HADHB; hydroxyacyl-Co; hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit; hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), beta aubunit; hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), beta subunit; mitochondrial trifunctional protein, beta subunit; MSTP029; MTPB; TP-BETA; trifunctional enzyme subunit beta, mitochondrial
Gene Symbols HADHB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HADHB. Recombinant protein control fragment (Product #RP-96092).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171155, 231086, 3032
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.