Learn More
Haemo-Sol™ Medical and Laboratory Instrument Cleaners
Click to view available options
Product Type:
Non-sudsing Cleaner
Regular Cleaner
Description
Keep items lasting longer and looking brand new!
- Used in laboratories, hospitals, medical, dental, and veterinary offices, as well as crime labs, environmental organizations, and pharmaceutical companies
- Simply soak items in Haemo-Sol and rinse – no need to scrub
- Products are 100% soluble and leave no soil or cleaner residue on parts or instruments
- Specially formulated to not rust metals or cloud glassware
Specifications
Specifications
| Product Type | Regular Cleaner |
| For Use With (Application) | Laboratories, hospitals, medical, dental, and veterinary offices, as well as by crime labs, electronic manufacturing plants, environmental organizations, and pharmaceutical companies |
| Volume (English) | 5 lb. |