Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haemophilus Influenzae B Antibody (1079/457), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064551AF532
Description
Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Haemophilus Influenzae B | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein G purified | |
Primary | |
Bacteria | |
IgG2a |
ELISA, Immunocytochemistry, Immunofluorescence | |
1079/457 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
H. influenza B | |
Native, NCTC 7279 | |
0.1 mL | |
NB100-64551 recognizes Haemophilus influenzae B, a gram-negative coccobacillus that is the cause of bacterial meningitis and other severe illnesses in young children. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction