Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haemophilus Influenzae B Antibody (1079/457), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064551FR
Description
Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Haemophilus Influenzae B | |
Monoclonal | |
DyLight 680 | |
H. influenza B | |
Native, NCTC 7279 | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
1079/457 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein G purified | |
RUO | |
NB100-64551 recognizes Haemophilus influenzae B, a gram-negative coccobacillus that is the cause of bacterial meningitis and other severe illnesses in young children. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction