Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haemophilus Influenzae B Antibody (1079/83), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064552C
Description
Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Haemophilus Influenzae B | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
NB100-64552 is reactive with Heamophilus Influenzae B. NB100-64552 is reactive with isolates fixed by phenol, formalin and boiling. | |
Store at 4C in the dark. | |
IgG2b |
ELISA, Immunocytochemistry, Immunofluorescence | |
1079/83 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
H. influenza B | |
Native, NCTC 7279 | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction