Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Haemophilus influenzae B Monoclonal Antibody (1079/457)
GREENER_CHOICE

Catalog No. MA183200
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183200 Supplier Invitrogen™ Supplier No. MA183200
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Haemophilus influenzae B antibody, clone 1079/457 recognizes Haemophilus influenzae B, a gram-negative coccobacillus that is the cause of bacterial meningitis and other severe illnesses in young children.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Haemophilus influenzae B
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1079/457
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias Haemophilus b; Hib; influenza b
Host Species Mouse
Immunogen Haemophilus influenzae B.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.