Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haemophilus influenzae LPS Antibody (12D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275002
Description
Haemophilus influenzae LPS Monoclonal specifically detects Haemophilus influenzae LPS in Bacteria samples. It is validated for N/A.Specifications
Haemophilus influenzae LPS | |
12D9 | |
Unconjugated | |
H. influenzae Lipopolysaccharide, H. influenzae LPS, Haemophilus influenzae Lipopolysaccharide | |
LPS epitope, TEPC-15 | |
0.1 mL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Monoclonal | |
1 mg/ml | |
PBS with 0.02% Sodium Azide | |
Mouse | |
Protein A purified | |
Immunology, Virology Bacteria and Parasites | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction