Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haemophilus influenzae P6 Antibody (7F3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP274999 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274999 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274999 Supplier Novus Biologicals Supplier No. NBP274999
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Haemophilus influenzae P6 Monoclonal antibody specifically detects Haemophilus influenzae P6 in Bacteria, Virus samples.

Specifications

Antigen Haemophilus influenzae P6
Classification Monoclonal
Clone 7F3
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Gene Alias Outer membrane protein P6
Host Species Mouse
Immunogen Conserved epitope on P6 protein
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria, Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.