Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium foil, 0.025mm (0.001in) thick, 99.5% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Catalog No. p-7050917
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]

Specifications

Quantity 50 x 50 mm
Odor Odorless
Weight ≈3.3g/100x100mm
Form Foil
Percent Purity 99.5%
Assay (metals basis excluding Zr), Zr nominal 3%
Chemical Name or Material Hafnium, 0.025mm (0.001 in.) thick
EINECSNumber 231-166-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.