Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%

Catalog No. AA1183414
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g

CAS: 13499-05-3 | Cl4Hf | 320.29 g/mol

Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13499-05-3
Molecular Formula Cl4Hf
Molecular Weight (g/mol) 320.29
MDL Number MFCD00011034
InChI Key PDPJQWYGJJBYLF-UHFFFAOYSA-J
Synonym hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis
PubChem CID 37715
IUPAC Name tetrachlorohafnium
SMILES Cl[Hf](Cl)(Cl)Cl

Specifications

Melting Point 320° (sublimation)
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range >98% (Metals basis excluding Zr)
Solubility Information Soluble in water,methanol and acetone.
Formula Weight 320.3
Odor Irritating
Sensitivity Moisture sensitive
Chemical Name or Material Hafnium(IV) chloride
Hazard Category H314
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 236-826-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.