Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium(IV) oxide, 99% (metals basis excluding Zr), Zr <1.5%, Thermo Scientific Chemicals

Catalog No. p-4412875
Click to view available options
Quantity:
25 g
100 g

Hafnium(IV) oxide is used in optical coatings and in advanced metal-oxide-semiconductor devices. It is used as a gate insulator in field-effect transistors due to its high dielectric constant. It plays an important role as a possible candidate for resistive-switching memories, as a refractory material in the insulation of devices such as thermocouples and as a high-κ dielectric in DRAM capacitors. Also, used in the preparation of hafnium tetrachloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12055-23-1
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Melting Point 2774°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (Metals Basis)
UN Number 3260
Solubility Information Insoluble in water.
Formula Weight 210.49
Density 9.68 g/mL
Chemical Name or Material Hafnium(IV) oxide
EINECSNumber 235-013-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.