Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAGH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528292 Supplier Invitrogen™ Supplier No. PA528292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, mouse liver. Predicted reactivity: Mouse (92%), Rat (90%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The enzyme encoded by the HAGH gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. Three transcript variants encoding different isoforms have been found for this gene. Diseases associated with HAGH include Hydroxyacyl Glutathione Hydrolase Deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAGH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene HAGH
Gene Accession No. Q16775, Q99KB8
Gene Alias BC019817; GLO2; Glo-2; Glx II; GLX2; GLXII; Glyoxalase II; glyoxylase 2; HAGH; HAGH1; hydroxyacyl glutathione hydrolase; hydroxyacylglutathione hydrolase; hydroxyacylglutathione hydrolase, mitochondrial; hydroxyacylglutathione hydroxylase; round spermatid protein RSP29; Rsp29
Gene Symbols HAGH
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HAGH. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14651, 3029
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.