Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAGH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580682 Supplier Invitrogen™ Supplier No. PA580682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human HAGH/GLO2/Glyoxalase II.

The enzyme encoded by the HAGH gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. Three transcript variants encoding different isoforms have been found for this gene. Diseases associated with HAGH include Hydroxyacyl Glutathione Hydrolase Deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAGH
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HAGH
Gene Accession No. Q16775
Gene Alias BC019817; GLO2; Glo-2; Glx II; GLX2; GLXII; Glyoxalase II; glyoxylase 2; HAGH; HAGH1; hydroxyacyl glutathione hydrolase; hydroxyacylglutathione hydrolase; hydroxyacylglutathione hydrolase, mitochondrial; hydroxyacylglutathione hydroxylase; round spermatid protein RSP29; Rsp29
Gene Symbols HAGH
Host Species Rabbit
Immunogen Recombinant Human HAGH/GLO2/Glyoxalase II protein (Met1-Asp260).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3029
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.