Learn More
Invitrogen™ HAGH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593097
Description
Immunogen sequence: MKVEVLPALT DNYMYLVIDD ETKEAAIVDP VQPQKVVDAA RKHGVKLTTV LTTHHHWDHA GGNEKLVKLE SGLKVYGGDD RIGALTHKIT HLSTLQVGSL NVKCLATPCH TSGHICYFVS KPGGSEPPAV FTGDTLFVAG CGKFYEGTAD EMCKALLEVL GRLPPDTRVY CGHEYTINNL KFARHVEPGN AAIREKLAWA KEKYSIGEPT VPSTLAEEFT YNPFMRVREK TVQQHAGETD PVTTMRAVRR EKDQFKMPRD; Positive Samples: MCF7, HepG2, Mouse liver, Mouse kidney, Mouse heart, Rat liver, Rat kidney; Cellular Location: Cytoplasm, Mitochondrion matrix.
The enzyme encoded by the HAGH gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. Three transcript variants encoding different isoforms have been found for this gene. Diseases associated with HAGH include Hydroxyacyl Glutathione Hydrolase Deficiency.
Specifications
HAGH | |
Polyclonal | |
Unconjugated | |
HAGH | |
BC019817; GLO2; Glo-2; Glx II; GLX2; GLXII; Glyoxalase II; glyoxylase 2; HAGH; HAGH1; hydroxyacyl glutathione hydrolase; hydroxyacylglutathione hydrolase; hydroxyacylglutathione hydrolase, mitochondrial; hydroxyacylglutathione hydroxylase; round spermatid protein RSP29; Rsp29 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14651, 24439, 3029 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35952, Q16775, Q99KB8 | |
HAGH | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human HAGH (NP_0010355171). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.