Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAGHL Antibody (OTI3H2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272283B
Description
HAGHL Monoclonal specifically detects HAGHL in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
HAGHL | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
HAGHL | |
Full length recombinant protein of human HAGHL (NP_115680) produced in HEK293T cells. | |
0.1 mL | |
84264.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI3H2 | |
Western Blot, Flow Cytometry | |
EC 3.1.2.-, GLO2-like/ RJD12, hydroxyacyl glutathione hydrolase-like, hydroxyacylglutathione hydrolase-like, hydroxyacylglutathione hydrolase-like protein, MGC2605 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction