Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAGHL Monoclonal Antibody (OTI1G8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502476S


Description
HAGHL Monoclonal antibody specifically detects HAGHL in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
HAGHL | |
Monoclonal | |
0.68 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q14CS0 | |
Mouse | |
Affinity chromatography | |
RUO | |
137886 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI1G8 | |
Unconjugated | |
UBXN2B | |
1500017E18Rik; 2810014I23Rik; C330022E15Rik; GLO2-like/ RJD12; Haghl; hydroxyacylglutathione hydrolase like; hydroxyacylglutathione hydrolase-like; hydroxyacylglutathione hydrolase-like protein | |
Full length human recombinant protein of human UBXN2B produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction