Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAGHL Monoclonal Antibody (OTI3H2)
GREENER_CHOICE

Catalog No. PIMA525448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525448 Supplier Invitrogen™ Supplier No. MA525448
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HAGHL Monoclonal Antibody for Western Blot, Flow

HAGHL is a hydrolase acting on ester bonds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAGHL
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3H2
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HAGHL
Gene Accession No. Q6PII5
Gene Alias 1500017E18Rik; 2810014I23Rik; C330022E15Rik; GLO2-like/ RJD12; Haghl; hydroxyacylglutathione hydrolase like; hydroxyacylglutathione hydrolase-like; hydroxyacylglutathione hydrolase-like protein
Gene Symbols HAGHL
Host Species Mouse
Immunogen Full length human recombinant protein of HAGHL produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84264
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.