Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAI-1/HGFA Inhibitor 1 Antibody (OTI4H2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271296AF405
Description
HAI-1/HGFA Inhibitor 1 Monoclonal specifically detects HAI-1/HGFA Inhibitor 1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
HAI-1/HGFA Inhibitor 1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SPINT1 | |
Full length recombinant protein of human HAI-1/HGFA Inhibitor 1 (NP_857593) produced in HEK293T cells. | |
0.1 mL | |
6692.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI4H2 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction