Learn More
HAI-1/HGFA Inhibitor 1 Antibody (RM0088-3G21), Alexa Fluor™ 350, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP212356AF350
Catalog No. NP21236A30
Only null left
Add to Cart
Description
Description
HAI-1/HGFA Inhibitor 1 Monoclonal specifically detects HAI-1/HGFA Inhibitor 1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
HAI-1/HGFA Inhibitor 1
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
SPINT1
Mouse recombinant HAI-1/HGFA Inhibitor 1 (extracellular domain)
0.1 mL
6692
Store at 4°C in the dark.
Western Blot
RM0088-3G21
Western Blot
HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1
Rat
Protein A or G purified
Primary
Mouse
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.