Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAI-1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547053
Description
In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse HAI-1 is observed and less than 1% cross-reactivity with recombinant human HAI-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.
Specifications
HAI-1 | |
Polyclonal | |
Unconjugated | |
SPINT1 | |
HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6692 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O43278 | |
SPINT1 | |
Mouse myeloma cell line NS0-derived recombinant human HAI-1 Pro37-Glu449. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction