Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAI-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117046 Supplier Invitrogen™ Supplier No. PA5117046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse liver, Rat brain Immunogen sequence: ANVTVTVLST KQTEDYCLAS NKVGRCRGSF PRWYYDPTEQ ICKSFVYGGC LGNKNNYLRE EECILACRGV QGPSMERRHP VCSGTCQPTQ FRCSNGCCID SFLECDDTPN CPDASDEAAC EKYTSGFDEL QRIHFPSDKG HCVDLPDTGL CKESIPRWYY NPFSEHCARF TYGGCYGNKN NFEEEQQCLE SCRGISKKDV FGLRREIPIP STGSVEMAVA VFLVICIVVV VAILGYCFFK NQRKDFHGHH HHPPPTPASS TVSTTEDTEH LVYNHTTRPL.

The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAI-1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPINT1
Gene Accession No. O43278, Q9R097
Gene Alias HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256
Gene Symbols SPINT1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20732, 311331, 6692
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.