Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAI-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142599
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142599 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142599 Supplier Invitrogen™ Supplier No. PA5142599
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize to recognize both reported isoforms (NP_066925.1; NP_001159575.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.

This gene encodes a transmembrane protein with two extracellular Kunitz domains that inhibits a variety of serine proteases. The protein inhibits HGF activator which prevents the formation of active hepatocyte growth factor. This gene is a putative tumor suppressor, and mutations in this gene result in congenital sodium diarrhea. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAI-2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SPINT2
Gene Accession No. O43291
Gene Alias AL024025; C76321; DIAR3; FLJ45571; HAI2; HAI-2; Hepatocyte growth factor activator inhibitor type 2; Kop; Kunitz-type protease inhibitor 2; PB; placental bikunin; serine peptidase inhibitor, Kunitz type 2; serine peptidase inhibitor, Kunitz type, 2; serine protease inhibitor, Kunitz type 2; serine protease inhibitor, Kunitz type, 2; Spint2; testicular tissue protein Li 183
Gene Symbols SPINT2
Host Species Goat
Immunogen Peptide with sequence C-SGDDKEQLVKNT.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10653
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.